Skip to Content
Merck
1-Hexene analytical standard

1-Hexene

1-Hexene analytical standard
Linear Formula:
CH3(CH2)3CH=CH2
CAS Number:
Molecular Weight:
84.16
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing