![1H,1H,2H,2H-Perfluorooctyl iodide 96%](/deepweb/assets/sigmaaldrich/product/structures/849/140/68d705a9-ed49-4681-92eb-75732f747166/640/68d705a9-ed49-4681-92eb-75732f747166.png)
1H,1H,2H,2H-Perfluorooctyl iodide
![1H,1H,2H,2H-Perfluorooctyl iodide 96%](/deepweb/assets/sigmaaldrich/product/structures/849/140/68d705a9-ed49-4681-92eb-75732f747166/640/68d705a9-ed49-4681-92eb-75732f747166.png)
Synonym(s):
1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodooctane, 1H,1H,2H,2H-Perfluorooctyl iodide, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl iodide, 8-Iodo-1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluorooctane
Linear Formula:
CF3(CF2)5CH2CH2I
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing