Skip to Content
Merck
1-Chlorocarbonyl-1-methylethyl acetate 95%

1-Chlorocarbonyl-1-methylethyl acetate

1-Chlorocarbonyl-1-methylethyl acetate 95%
Synonym(s):
α-Acetoxyisobutyryl chloride, 2-Acetoxy-2-methylpropionyl chloride
Linear Formula:
(CH3)2C(OCOCH3)COCl
CAS Number:
Molecular Weight:
164.59
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing