Skip to Content
Merck
1-Bromo-2-difluoromethylbenzene 97%

1-Bromo-2-difluoromethylbenzene

1-Bromo-2-difluoromethylbenzene 97%
Synonym(s):
2-Bromo-1-(difluoromethyl)benzene
Empirical Formula (Hill Notation):
C7H5BrF2
CAS Number:
Molecular Weight:
207.02

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing