Skip to Content
Merck
1,4-Cubanedicarboxylic acid ≥97%

1,4-Cubanedicarboxylic acid

1,4-Cubanedicarboxylic acid ≥97%
Empirical Formula (Hill Notation):
C10H8O4
CAS Number:
Molecular Weight:
192.17

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing