Skip to Content
Merck
1,4-Bis(2-methylstyryl)benzene BioReagent, suitable for scintillation, ≥98% (UV)

1,4-Bis(2-methylstyryl)benzene

1,4-Bis(2-methylstyryl)benzene BioReagent, suitable for scintillation, ≥98% (UV)
Synonym(s):
Bis-MSB
Linear Formula:
(CH3C6H4CH=CH)2C6H4
CAS Number:
Molecular Weight:
310.43
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Special Grade
Showing 1-1 of 1