![1,3,5-O-Methylidyne-myo-inositol ≥99.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/179/638/31ac58c4-7dd6-40ca-9280-d755a0b4bfb9/640/31ac58c4-7dd6-40ca-9280-d755a0b4bfb9.png)
1,3,5-O-Methylidyne-myo-inositol
![1,3,5-O-Methylidyne-myo-inositol ≥99.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/179/638/31ac58c4-7dd6-40ca-9280-d755a0b4bfb9/640/31ac58c4-7dd6-40ca-9280-d755a0b4bfb9.png)
Synonym(s):
myo-Inositol monoorthoformate
Empirical Formula (Hill Notation):
C7H10O6
CAS Number:
Molecular Weight:
190.15
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing