1,2-dipiperidinoethane
Empirical Formula (Hill Notation):
C12H24N2
CAS Number:
Molecular Weight:
196.33
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing