1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine
Synonym(s):
(9Z)-9-Octadecenoic acid (1R)-1-[[[(2-aminoethoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl ester, 1,2-Di(9Z-octadecenoyl)-sn-glycero-3-phosphoethanolamine, 3-sn-Phosphatidylethanolamine, 1,2-dideoyl, L-β,γ-Dioleoyl-α-cephalin, DOPE, Dioleoyl phosphatidylethanolamine, PE(18:1(9Z)/18:1(9Z))
Empirical Formula (Hill Notation):
C41H78NO8P
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Functional Group
Optical Isomer
Showing 1-2 of 2