Skip to Content
Merck
1,2-Dilinoleoyl-3-oleoyl-rac-glycerol ≥98%

1,2-Dilinoleoyl-3-oleoyl-rac-glycerol

1,2-Dilinoleoyl-3-oleoyl-rac-glycerol ≥98%
Synonym(s):
1,2-Di(cis-9,12-octadecadienoyl)-3-(cis-9-octadecenoyl)-rac-glycerol, OLL
Empirical Formula (Hill Notation):
C57H100O6
CAS Number:
Molecular Weight:
881.40

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing