Skip to Content
Merck
(−)-1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene kanata purity

(−)-1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene

(−)-1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene kanata purity
Synonym(s):
(2R,2′R,5R,5′R)-2,2′-5,5′-Tetraethyl-1,1′-(o-phenylene)diphospholane, (R,R)-Ethyl-DUPHOS
Empirical Formula (Hill Notation):
C22H36P2
CAS Number:
Molecular Weight:
362.47

Products

Brand
Formula Weight
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing