Skip to Content
Merck
1,2,6-Hexanetriol 96%

1,2,6-Hexanetriol

1,2,6-Hexanetriol 96%
Synonym(s):
1,2,6-Trihydroxyhexane, 1,2,6-Hexanetriol
Linear Formula:
HO(CH2)4CH(OH)CH2OH
CAS Number:
Molecular Weight:
134.17

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing