![11-Keto-β-boswellic acid analytical standard](/deepweb/assets/sigmaaldrich/product/structures/204/013/6e63b4cb-3373-4c80-aba8-b2fcfcbf28fb/640/6e63b4cb-3373-4c80-aba8-b2fcfcbf28fb.png)
11-Keto-β-boswellic acid
![11-Keto-β-boswellic acid analytical standard](/deepweb/assets/sigmaaldrich/product/structures/204/013/6e63b4cb-3373-4c80-aba8-b2fcfcbf28fb/640/6e63b4cb-3373-4c80-aba8-b2fcfcbf28fb.png)
Synonym(s):
11-Oxo-β-boswellic acid, 3α-Hydroxy-11-oxours-12-en-24-oic acid, KBA
Empirical Formula (Hill Notation):
C30H46O4
CAS Number:
Molecular Weight:
470.68
Products
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing