
1,1′-Dioctadecyl-4,4′-bipyridinium dibromide

Synonym(s):
Octadecyl viologen dibromide
Empirical Formula (Hill Notation):
C46H82Br2N2
CAS Number:
Molecular Weight:
822.96
Products
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Showing 1-1 of 1