Skip to Content
Merck
1,12-Dodecanediol 99%

1,12-Dodecanediol

1,12-Dodecanediol 99%
Synonym(s):
Dodecamethylene glycol, 1,12-Dodecanediol
Linear Formula:
HO(CH2)12OH
CAS Number:
Molecular Weight:
202.33

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing