Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate
Synonym(s):
β,β,β-Trichloro-
t
-butanol, β,β,β-Trichloro-
tert
-butyl alcohol hemihydrate, Acetone chloroform, Chloretone, Chlorobutanol, ‘Acetone chloroform’, ‘Chlorobutanol’
Linear Formula:
Cl
3
CC(CH
3
)
2
OH · 0.5H
2
O
CAS Number:
6001-64-5
Molecular Weight:
186.46
EC Number:
249-042-3
Products
Product Category
building blocks (1)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
112054
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate,
98%
Expand
View Pricing
20801
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate,
meets analytical specification of Ph. Eur., BP, NF, 98-100.5% (calc with ref. to anhyd. subst.)
Expand
View Pricing