Product Name
Methyl 5-(4-aminophenyl)furan-2-carboxylate 97%
Product Number
648469
Product Brand
aldrich
CAS Number
52939-06-7
Molecular Weight
217.22
Test | Specification |
---|---|
Appearance (Color) | White to Tan |
Appearance (Form) | Conforms to Requirement |
Powder or Crystals | |
Infrared spectrum | Conforms to Structure |
Carbon | 64.0 - 68.7 % |
Nitrogen | 6.2 - 6.7 % |
Purity (GC) | ≥96.5 % |