Skip to Content
Merck
All Photos(3)

Key Documents

V900654

Sigma-Aldrich

2,3-Diaminopyridine

Vetec, reagent grade, 94%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7N3
CAS Number:
Molecular Weight:
109.13
Beilstein:
109869
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

94%

mp

110-115 °C (lit.)

SMILES string

Nc1cccnc1N

InChI

1S/C5H7N3/c6-4-2-1-3-8-5(4)7/h1-3H,6H2,(H2,7,8)

InChI key

ZZYXNRREDYWPLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ismail Yildirim et al.
Molecules (Basel, Switzerland), 10(3), 559-571 (2007-11-17)
The 1H-pyrazole-3-carboxylic acid 2 was converted in good yield (69%) into the corresponding 1H-pyrazole-3-carboxamide 5 via reaction of the acid chloride 3 with 2,3- diaminopyridine (4). A different product, the 3H-imidazo[4,5-b] pyridine derivative 6, was formed from the reaction of
G E Kirsch et al.
The Journal of pharmacology and experimental therapeutics, 226(1), 174-179 (1983-07-01)
Aminopyridines, potent potassium channel blocking agents, were studied for their site of action and active form in the nerve membrane. Voltage clamped, internally perfused squid giant axons were used. 3,4-Diaminopyridine, one of the most potent aminopyridine derivatives, blocked the potassium
Fengli Yu et al.
Talanta, 78(4-5), 1395-1400 (2009-04-14)
A new voltammetric enzyme-linked immunoassay system using the electrochemical substrate 2,3-diaminopyridine (DAP) and horseradish peroxidase (HRP) system has been developed. DAP is oxidized with H2O2 catalyzed by HRP, and the resulting electroactive product produces a sensitive voltammetric peak at potential
Narjes Deqnah et al.
Anticancer research, 32(12), 5331-5336 (2012-12-12)
In this article, we report the synthesis and the in vitro activity of trans-bis(2-methylimidazole)dichloroplatinum(II) (coded as DH4) and trans-(ammino)(2,3-diaminopyridine) dichloroplatinum(II) (coded as DH5) in the human ovarian tumour models. DH4 is less active than cisplatin against the parental A2780 cell
Nessreen A Al-Hashimi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 75(1), 198-202 (2009-11-26)
The charge transfer (CT) interaction between iodine and 2,3-diaminopyridine (DAPY) has been thoroughly investigated via theoretical calculations. A Hartree-Fock, 3-21G level of theory was used to optimize and calculate the Mullican charge distribution scheme as well as the vibrational frequencies

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service