Skip to Content
Merck
All Photos(1)

Key Documents

TA9H97BAF25D

Targetmol Chemicals Inc.

TB500 acetate(885340-08-9 free base)

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
949.07

Iupac Name

(2S,5S,8S,11S,14S,17S,20S)-2-(3-amino-3-oxopropyl)-14,17-bis(4-aminobutyl)-8-(2-carboxyethyl)-5,11-bis((R)-1-hydroxyethyl)-20-isobutyl-4,7,10,13,16,19,22-heptaoxo-3,6,9,12,15,18,21-heptaazatricosanoic acid--acetic acid (1/1)

Inchi Code

1S/C38H68N10O14.C2H4O2/c1-19(2)18-27(42-22(5)51)35(58)44-23(10-6-8-16-39)32(55)43-24(11-7-9-17-40)33(56)47-30(20(3)49)36(59)45-25(13-15-29(53)54)34(57)48-31(21(4)50)37(60)46-26(38(61)62)12-14-28(41)52;1-2(3)4/h19-21,23-27,30-31,49-50H,6-18,39-40H2,1-5H3,(H2,41,52)(H,42,51)(H,43,55)(H,44,58)(H,45,59)(H,46,60)(H,47,56)(H,48,57)(H,53,54)(H,61,62);1H3,(H,3,4)/t20-,21-,23+,24+,25+,26+,27+,30+,31+;/m1./s1

InChI key

QQPHIKZJJGWYIR-PFWNPZQHSA-N

storage temp.

-80/-20

Purity

100%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/TB500 acetate(885340-08-9 free base)

MSDS

https://www.targetmol.com/compound/TB500 acetate(885340-08-9 free base)


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service