Skip to Content
Merck
All Photos(1)

Key Documents

SY3H97AC690B

Synthonix Corporation

7-Bromo-3,4-dihydroquinazolin-4-one

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
HUF 178,144.00
250 MG
HUF 246,316.00
500 MG
HUF 378,214.00
1 G
HUF 480,472.00
2.5 G
HUF 925,072.00
5 G
HUF 1,361,274.00
10 G
HUF 2,010,390.00

Vendor SKU: EN300-191825-100MG

HUF 178,144.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
HUF 178,144.00
250 MG
HUF 246,316.00
500 MG
HUF 378,214.00
1 G
HUF 480,472.00
2.5 G
HUF 925,072.00
5 G
HUF 1,361,274.00
10 G
HUF 2,010,390.00

About This Item

CAS Number:
Molecular Weight:
225.04
MDL number:

Vendor SKU: EN300-191825-100MG

HUF 178,144.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

7-bromoquinazolin-4(1H)-one

Inchi Code

1S/C8H5BrN2O/c9-5-1-2-6-7(3-5)10-4-11-8(6)12/h1-4H,(H,10,11,12)

InChI key

FUGKKMYSCAYXJJ-UHFFFAOYSA-N

storage temp.

Room Temperature

Purity

97%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Solid

Product Link

https://synthonix.com/1649.html

Pictograms

Exclamation mark

Signal Word

Warning

Precautionary statements

MSDS

https://synthonix.com/1649.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service