SY3H3D67F650
Synthonix Corporation
Methyl 2-(chlorosulfonyl)acetate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl (chlorosulfonyl)acetate
Inchi Code
1S/C3H5ClO4S/c1-8-3(5)2-9(4,6)7/h2H2,1H3
InChI key
YBIPZPBGAGTBGK-UHFFFAOYSA-N
storage temp.
Refrigerator
Purity
97%
Country of Origin
US
Shipping Temperature
Room Temperature
Physical Form
Liquid
Product Link
https://synthonix.com/39862.html
Signal Word
Danger
Hazard Statements
MSDS
https://synthonix.com/39862.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service