Journal of separation science, 29(13), 1977-1987 (2006-10-05)
A well established method of direct injection of larger than conventional sample volumes ranging from 0.1 mL to 10 mL in HPLC is the injection valve method in which a loop of tubing is totally or partially filled with sample.
Journal of environmental quality, 36(3), 826-831 (2007-05-09)
Mineralization of atrazine and formation of extractable and non-extractable "bound" residues were followed under laboratory conditions in two contrasting soils (organic C, texture, and atrazine application history) from northern Spain. The soils, a Humic Cambisol (MP) and a Gleyic Cambisol
Toxicological sciences : an official journal of the Society of Toxicology, 76(1), 190-200 (2003-09-13)
We showed previously that the chlorotriazine herbicide, atrazine (ATR), delays the onset of pubertal development in female rats. ATR and its biotransformation by-products are present in soil and groundwater. Since current maximum contaminant levels are set only for ATR, it
Soil leaching column chromatographic technique for estimation of leaching behavior of atrazine, deethylatrazine, deisopropylatrazine, and hydroxyatrazine on soil.
F Xu et al.
Bulletin of environmental contamination and toxicology, 63(1), 87-93 (1999-07-29)
Toxicological sciences : an official journal of the Society of Toxicology, 59(1), 127-137 (2001-01-03)
The effects of four major chlorotriazine metabolites on the constitutive synthesis of the catecholamines dopamine (DA) and norepinephrine (NE) were examined, using undifferentiated PC12 cells. NE release and intracellular DA and NE concentrations were quantified, for up to 24 h
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.