Skip to Content
Merck
All Photos(1)

Key Documents

Z605190

Eppendorf® ThermoStat Plus

w/o interchangeable block, AC/DC input 100 / 240 V AC, 50-60 Hz

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

manufacturer/tradename

Eppendorf® 5352 000.010 (European)
Eppendorf® 5352 000.037 (UK)

availability

not available in USA and Canada

W × D × H

22 cm × 25 cm × 12 cm

Looking for similar products? Visit Product Comparison Guide

General description

ThermoStat Plus controls temperature at your lab bench, accurately and conveniently. This compact, benchtop thermostat maintains different temperature levels up to 99°C, with a cooling feature for temperatures as low as -5°C.

Applications include: Enzyme storage at 0°C, Enzyme reactions at 37°C, Reactions at temperatures typically between 12 and 65°C.

  • Temperature and run times are individually programmable
  • Stores up to four temperatures and times in succession
  • RS-232 interface-provides control and allows documentation in accordance with GLP
  • Temperature Control Range of Block from -5 to 99°C
  • Range of Possible Temperatures from 30°C below ambient to 99°C
  • Minimum Block Temperature: -25°C at ambient of 4°C
  • Accuracy of Temperature Control: ±1°C between -5°C and 45°C; ±2°C between 45°C and 99°C
  • Heating-up Speed: 4°C/min
  • Cooling-down Speed: 2°C/min between 99°C and 25°C; 1°C/min between 25°C and -5°C
  • Cooling Time: from 22 to 0°C in 20 minutes
  • Range of Time Phases: 0:01 to 99:59 hours
  • Dimensions, without block (W x D x H): 8.7 x 9.8 x 4.9 in/22 x 25 x 12.5 cm
ThermoStat Plus controls temperature at your lab bench, accurately and conveniently. This compact, benchtop thermostat maintains different temperature levels up to 99°C, with a cooling feature for temperatures as low as -5°C
  • Temperature and run times are individually programmable
  • RS-232 interface-provides control and allows documentation in accordance with GLP
  • Range of Possible Temperatures from 30°C below ambient to 99°C

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service