Skip to Content
Merck
All Photos(1)

Key Documents

X2754

Sigma-Aldrich

Xanomeline L-tartrate hydrate

≥98% (HPLC)

Synonym(s):

3-Hexoxy-4-(1-methyl-3,6-dihydro-2H-pyridin-5-yl)-1,2,5-thiadiazole, LY-246708, Lomeron, Memcor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H23N3OS · xC4H6O6 · yH2O
Molecular Weight:
281.42 (anhydrous free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated
under inert gas

color

off-white

solubility

H2O: 10 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CCCCCCOc1nsnc1C2=CCCN(C)C2

InChI

1S/C14H23N3OS.C4H6O6.H2O/c1-3-4-5-6-10-18-14-13(15-19-16-14)12-8-7-9-17(2)11-12;5-1(3(7)8)2(6)4(9)10;/h8H,3-7,9-11H2,1-2H3;1-2,5-6H,(H,7,8)(H,9,10);1H2/t;1-,2-;/m.1./s1

InChI key

YTWKANAWFZNIKV-UACZKNEHSA-N

Biochem/physiol Actions

Xanomeline L-tartate is a M1 muscarinic receptor agonist.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristine B Jørgensen et al.
Journal of comparative physiology. A, Neuroethology, sensory, neural, and behavioral physiology, 202(1), 7-15 (2015-11-02)
The present study investigated the cholinergic system in the African naked mole-rat (Heterocephalus glaber) with focus on the muscarinic acetylcholine receptor subtypes M1 and M4. The protein sequences for the subtypes m 1-5 of the naked mole-rat were compared to
Karin Hellman et al.
ACS chemical neuroscience, 11(2), 173-183 (2019-12-19)
Current antipsychotic drugs are notably ineffective at addressing the cognitive deficits associated with schizophrenia. N-Desmethylclozapine (NDMC), the major metabolite of clozapine, displays muscarinic M1 receptor (M1) agonism, an activity associated with improvement in cognitive functioning. Preclinical and clinical data support

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service