Skip to Content
Merck
All Photos(4)

Key Documents

WH0000246M4

Sigma-Aldrich

Monoclonal Anti-ALOX15 antibody produced in mouse

clone 3D8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-arachidonate 15-lipoxygenase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3D8, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALOX15(246)

General description

The gene arachidonate 15-lipoxygenase (ALOX15) is mapped to human chromosome 17p13. It is mainly present in the epithelial cells in the upper airways, reticulocytes, eosinophils and macrophages. ALOX15 belongs to the dioxygenases family. It is an IL-4 (interleukin 4)/IL-13 target gene.

Immunogen

ALOX15 (AAH29032, 1 a.a. ~ 662 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MGLYRIRVSTGASLYAGSNNQVQLWLVGQHGEAALGKRLWPARGKETELKVEVPEYLGPLLFVKLRKRHLLKDDAWFCNWISVQGPGAGDEVRFPCYRWVEGNGVLSLPEGTGRTVGEDPQGLFQKHREEELEERRKLYRWGNWKDGLILNMAGAKLYDLPVDERFLEDKRVDFEVSLAKGLADLAIKDSLNVLTCWKDLDDFNRIFWCGQSKLAERVRDSWKEDALFGYQFLNGANPVVLRRSAHLPARLVFPPGMEELQAQLEKELEGGTLFEADFSLLDGIKANVILCSQQHLAAPLVMLKLQPDGKLLPMVIQLQLPRTGSPPPPLFLPTDPPMAWLLAKCWVRSSDFQLHELQSHLLRGHLMAEVIVVATMRCLPSIHPIFKLIIPHLRYTLEINVRARTGLVSDMGIFDQIMSTGGGGHVQLLKQAGAFLTYSSFCPPDDLADRGLLGVKSSFYPQDALRLWEIIYRYVEGIVSLHYKTDVAVKDDPELQTWCREITEIGLQGAQDRGFPVSLQARDQVCHFVTMCIFTCTGQHASVHLGQLDWYSWVPNAPCTMRLPPPTTKDATLETVMATLPNFHQASLQMSITWQLGRRQPVMVAVGQHEEEYFSGPEPKAVLKKFREELAALDKEIEIRNAKLDMPYEYLRPSVVENSVAI

Biochem/physiol Actions

ALOX15 (arachidonate 15-lipoxygenase) is involved in homeostatic as well as pathological responses. ALOX15 is responsible for the oxidation of unsaturated fatty acids (at the 15 position) to active hydroperoxy and epoxy metabolites. It is also responsible for the conversion of linoleic acid to anti-tumor 13-S-hydroxyoctadecadienoic acid. ALOX15 is required for IL-4 (interleukin-4)/IL-13-mediated macrophage polarization. In mice, it is a negative regulator of bone mineral density. It is also involved in fatty acid metabolism. It controls MAPK (mitogen activated protein kinase) signaling in human airway epithelial cells using phosphatidylethanolamine-binding protein.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

12/15 Lipoxygenase regulation of colorectal tumorigenesis is determined by the relative tumor levels of its metabolite 12-HETE and 13-HODE in animal models.
Chang J, et al.
Oncotarget, 6, 2879-2879 (2015)
Polymorphisms in inflammation associated genes ALOX15 and IL-6 are associated with bone properties in young women and fracture in elderly.
Herlin M, et al.
Bone, 79, 105-105 (2015)
15-Lipoxygenase 1 interacts with phosphatidylethanolamine-binding protein to regulate MAPK signaling in human airway epithelial cells.
Zhao J, et al.
Proceedings of the National Academy of Sciences of the USA, 108, 14246-14246 (2011)
Interaction of human 15-lipoxygenase-1 with phosphatidylinositol bisphosphates results in increased enzyme activity.
Andersson E, et al.
Biochimica et Biophysica Acta, 1761, 1498-1498 (2006)
Dmitry Namgaladze et al.
The Journal of biological chemistry, 290(40), 24484-24494 (2015-08-16)
Macrophages respond to the Th2 cytokine IL-4 with elevated expression of arachidonate 15-lipoxygenase (ALOX15). Although IL-4 signaling elicits anti-inflammatory responses, 15-lipoxygenase may either support or inhibit inflammatory processes in a context-dependent manner. AMP-activated protein kinase (AMPK) is a metabolic sensor/regulator

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service