Skip to Content
Merck
All Photos(1)

Key Documents

SRP5165

Sigma-Aldrich

BAG1 (72-end), GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

RAP46

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$321.00

$321.00


Estimated to ship on19 April 2025


Request a Bulk Order

Select a Size

Change View
50 μG
$321.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$321.00


Estimated to ship on19 April 2025


Request a Bulk Order

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~66 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BAG1(573)

General description

BAG1 (also known as BCL2-associated athanogene) is a membrane protein rich in glutamic acid residues that binds to BCL2 and blocks apoptosis or programmed cell death. The BAG1-BCL2 complex enhances the anti-apoptotic effects of BCL2 and represents a link between growth factor receptors and anti-apoptotic mechanisms. Overexpression of BAG1 in 3T3 fibroblasts prevents apoptosis in the presence of low serum. BAG1 has also been shown to interact with activated glucocorticoid, androgen, estrogen and progesterone receptors. Binding to these receptors by BAG1 is dependent on receptor activation.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N K Clemo et al.
Biochemical Society transactions, 33(Pt 4), 676-678 (2005-07-27)
Although the retinoblastoma susceptibility gene RB1 is inactivated in a wide variety of human cancers, the retinoblastoma protein (Rb) has been shown to be overexpressed in colon cancers, which is linked to the anti-apoptotic function of the protein. However, the
Shou-Ching Tang
IUBMB life, 53(2), 99-105 (2002-06-07)
BAG-1 is a multifunctional and anti-apoptotic or anti-cell death protein that interacts with a variety of cellular proteins and affects their functions. On the cell surface, it binds to the cytosolic domain of the growth factor receptors and enhances the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service