Skip to Content
Merck
All Photos(1)

Documents

SRP5158

Sigma-Aldrich

14-3-3 θ, untagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

14-3-3, 14-3-3 θ, 1C5, HS1, YWHAQ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~29 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... YWHAQ(10971)

General description

14-3-3θ (also known as tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, theta polypeptide) is a member of the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. Through interaction with ASK1, c-jun NH-terminal kinase, and p38 mitogen-activated protein kinase (MAPK), 14-3-3θ plays an important role in controlling apotopsis. Induced expression of 14-3-3θ protein has been reported in patients with amyotrophic lateral sclerosis. Additionally, 14-3-3θ has been observed to mediate nucleocytoplasmic shuttling of the N protein (coronavirus nucleocapsid protein) which causes severe acute respiratory syndrome.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Milan Surjit et al.
Journal of virology, 79(17), 11476-11486 (2005-08-17)
The severe acute respiratory syndrome coronavirus(SARS-CoV) nucleocapsid (N) protein is one of the four structural proteins of the virus and is predicted to be a 46-kDa phosphoprotein. Our in silico analysis predicted N to be heavily phosphorylated at multiple residues.
Jeffrey M C Lau et al.
Molecular and cellular biology, 27(4), 1455-1466 (2006-12-06)
14-3-3 family members are intracellular dimeric phosphoserine-binding proteins that regulate signal transduction, cell cycle, apoptotic, and metabolic cascades. Previous work with global 14-3-3 protein inhibitors suggested that these proteins play a critical role in antagonizing apoptotic cell death in response

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service