Skip to Content
Merck
All Photos(1)

Documents

SML1026

Sigma-Aldrich

Lasofoxifene tartrate

≥98% (HPLC)

Synonym(s):

(-)-cis-6-Phenyl-5-[4-(2-pyrrolidin-1-ylethoxy)phenyl]-5,6,7,8-tetrahydronaphthalen-2-ol tartrate, (5R,6S)-5,6,7,8-Tetrahydro-6-phenyl-5-[4-[2-(1-pyrrolidinyl)ethoxy]phenyl]-2-naphthalenol tartrate, CP-336,156 tartrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H31NO2 · C4H6O6
CAS Number:
Molecular Weight:
563.64
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=CC=C(C(CC[C@@H]2C3=CC=CC=C3)=C1)[C@H]2C4=CC=C(OCCN5CCCC5)C=C4.O=C(O)[C@@H](O)[C@H](O)C(O)=O

InChI

1S/C28H31NO2.C4H6O6/c30-24-11-15-27-23(20-24)10-14-26(21-6-2-1-3-7-21)28(27)22-8-12-25(13-9-22)31-19-18-29-16-4-5-17-29;5-1(3(7)8)2(6)4(9)10/h1-3,6-9,11-13,15,20,26,28,30H,4-5,10,14,16-19H2;1-2,5-6H,(H,7,8)(H,9,10)/t26-,28+;1-,2-/m10/s1

InChI key

INEHJXCWEVNEDZ-LUDNRVPPSA-N

Biochem/physiol Actions

Lasofoxifene is a third-generation selective estrogen receptor modulator (SERM).
Lasofoxifene possesses strong resistance to intestinal wall glucuronidation and thus promotes oral bioavailability. In postmenopausal women, it reduces bone resorption, bone loss and low density lipoprotein (LDL) cholesterol. Selective and regular dose of Lasofoxifene is known to reduce the risk of fractures, breast cancer, major coronary heart disease, and stroke.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lasofoxifene in postmenopausal women with osteoporosis.
Cummings S R, et al.
The New England Journal of Medicine, 362(8), 686-696 (2010)
Lasofoxifene: a third-generation selective estrogen receptor modulator for the prevention and treatment of osteoporosis.
Gennari L, et al.
Expert Opinion on Investigational Drugs, 15(9), 1091-1103 (2006)
Nam Sook Kim et al.
Journal of separation science, 43(14), 2750-2765 (2020-04-17)
An analytical method for the simultaneous and reliable determination of 20 antigout and antiosteoporosis pharmaceutical compounds in adulterated health food products was developed using liquid chromatography with electrospray ionization tandem mass spectrometry and liquid chromatography with quadrupole-time-of-flight mass spectrometry. The
Kaitlyn J Andreano et al.
Breast cancer research and treatment, 180(3), 635-646 (2020-03-05)
The combination of targeting the CDK4/6 and estrogen receptor (ER) signaling pathways with palbociclib and fulvestrant is a proven therapeutic strategy for the treatment of ER-positive breast cancer. However, the poor physicochemical properties of fulvestrant require monthly intramuscular injections to
Nam Sook Kim et al.
Journal of pharmaceutical and biomedical analysis, 179, 113003-113003 (2019-12-10)
The aim of this study was to simultaneously determine the presence of unauthorized drug substances in health foods and herbal products used in the treatment of conditions such as gout and anti-osteoporosis. Therefore, we developed and optimised a rapid and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service