Skip to Content
Merck
All Photos(1)

Key Documents

SEP080

Millipore

Seppro® Rubisco

LC2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

column D × L

6.4 mm × 63 mm

capacity

1 mg total protein loading(D-Ribulose 1,5-Diphosphate Carboxylase (Rubisco))

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

RuBisCO (Ribulose-1,5-bisphosphate carboxylase/oxygenase) is the most abundant protein in plants, and it may be the most abundant protein on Earth. RuBisCO is an enzyme that is used in the Calvin cycle to catalyze the first major step of carbon fixation, a process by which the atoms of atmospheric carbon dioxide are made available to organisms in the form of energy-rich molecules such as sucrose. RuBisCO, while being the key enzyme in photosynthetic carbon assimilation in green leaves, is the main obstacle in plant proteomics. It constitutes about 40% of the total protein mass in green leaves, thus interfering with proteomics studies such as LC-MS/MS and 2D-gel electrophoresis. To meet the needs for specifically and effectively separating RuBisCO from other plant proteins, Sigma developed a novel immunoaffinity matrix based on the IgY antibodies cross-linked to microbeads, which specifically removes RuBisCO protein from plant extract.

Application

Seppro® Rubisco, LC2 (2mL columns) may be used in plant proteomics research to effect IgY-mediated immune affinity removal of RuBisCO (Ribulose-1,5-bisphosphate carboxylase/oxygenase) from plant tissue extracts via liquid chromatography.

Legal Information

Seppro is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Also Available Separately

Product No.
Description
SDS

  • S4199Seppro® Dilution bufferSDS

  • S4324Seppro® Stripping BufferSDS

  • S4449Seppro® Neutralization BufferSDS

  • CLS8163Corning® Costar® Spin-X® centrifuge tube filters, cellulose acetate membrane, pore size 0.45 μm, non-sterileSDS

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renato Millioni et al.
PloS one, 6(5), e19603-e19603 (2011-05-17)
To date, the complexity of the plasma proteome exceeds the analytical capacity of conventional approaches to isolate lower abundance proteins that may prove to be informative biomarkers. Only complex multistep separation strategies have been able to detect a substantial number

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service