Skip to Content
Merck
All Photos(1)

Key Documents

SAB4501909

Sigma-Aldrich

Anti-MAT1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

AMS1, AdoMet synthetase, MAT-I/III, MATA1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$589.00

$589.00


Estimated to ship on18 April 2025



Select a Size

Change View
100 μG
$589.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$589.00


Estimated to ship on18 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 35 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MNAT1(4331)

General description

Anti-MAT1 Antibody detects endogenous levels of total MAT1 protein. CDK activating kinase assembly factor MNAT1 or MAT1 enzyme is encoded by the MNAT1 gene in humans and is localized to chromosome 14q23. The protein is part of the cyclin-dependent kinase-activating kinase complex.

Immunogen

The antiserum was produced against synthesized peptide derived from human MAT1.

Immunogen Range: 91-140

Biochem/physiol Actions

CDK activating kinase assembly factor MNAT1 or MAT1 is functionally associated with the general transcription/DNA repair factor TFIIH. It is an assembly factor that targets subunits of both cyclin-dependent kinase-activating kinase (CAK) and TFIIH kinase. MAT1 regulates cell cycle and transcription. This protein associates with cyclin H and cyclin-dependent kinase 7 (CDK7) and forms the CDK-activating kinase (CAK) complex. The complex activates several cyclin-associated kinases, regulates cell cycle progression and activates RNA polymerase II-mediated transcription.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Retinoid-induced G1 arrest and differentiation activation are associated with a switch to cyclin-dependent kinase-activating kinase hypophosphorylation of retinoic acid receptor alpha
Jiwei Wang
The Journal of Biological Chemistry, 277 (2002)
Solution structure of the N-terminal domain of the human TFIIH MAT1 subunit: new insights into the RING finger family
V Gervais
The Journal of Biological Chemistry, 276 (2000)
The lost intrinsic fragmentation of MAT1 protein during granulopoiesis promotes the growth and metastasis of leukemic myeloblasts
Siyue Lou
Stem Cells (2013)
Mapping of the human genes encoding cyclin H (CCNH) and the CDK-activating kinase (CAK) assembly factor MAT1 (MNAT1) to chromosome bands 5q13.3-q14 and 14q23, respectively
Eki T
Genomics, 47(1) (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service