Skip to Content
Merck
All Photos(3)

Key Documents

SAB4500103

Sigma-Aldrich

Anti-TACC3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ERIC-1, ERIC1, TAC3, Transforming acidic coiled-coil-containing protein 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$589.00

$589.00


In StockDetails



Select a Size

Change View
100 μG
$589.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$589.00


In StockDetails


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 90 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TACC3(10460)

General description

Anti-TACC3 Antibody detects endogenous levels of total TACC3 protein.
Transforming acidic coiled-coil-containing protein 3 is encoded by TACC3 gene in humans and belonging to the human transforming acidic coiled-coil (TACC) family of centrosomal adaptor proteins. It is also known as ERIC1 and ERIC-1.

Immunogen

The antiserum was produced against synthesized peptide derived from human TACC3.

Immunogen Range: 789-838

Biochem/physiol Actions

Transforming acidic coiled-coil-containing protein 3 (TACC3) plays a critical role in microtubule nucleation at the centrosome. It is involved in the regulation of microtubule nucleation at the centrosome and functions in the stabilization of the γ-tubulin ring complex assembly. It plays an essential role in spindle assembly and centrosome integrity during mitosis as well as for cellular survival. It may act as a potential therapeutic target in cancer cells. TACC3 is aberrantly expressed in a variety of human cancers. It acts as a driver of tumorigenesis as well as an inducer of oncogenic EMT.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katelyn N Nelson et al.
Molecular cancer research : MCR, 14(5), 458-469 (2016-02-13)
Fibroblast growth factor receptors (FGFR) are critical for cell proliferation and differentiation. Mutation and/or translocation of FGFRs lead to aberrant signaling that often results in developmental syndromes or cancer growth. As sequencing of human tumors becomes more frequent, so does
Clark G Wang et al.
Oncotarget, 14, 133-145 (2023-02-14)
FGFR3-TACC3 represents an oncogenic fusion protein frequently identified in glioblastoma, lung cancer, bladder cancer, oral cancer, head and neck squamous cell carcinoma, gallbladder cancer, and cervical cancer. Various exon breakpoints of FGFR3-TACC3 have been identified in cancers; these were analyzed
A small compound targeting TACC3 revealed its different spatiotemporal contributions for spindle assembly in cancer cells.
Yao R, Kondoh Y, Natsume Y, et al.
Oncogene, 33(33), 4242-4252 (2014)
Puja Singh et al.
The Journal of biological chemistry, 289(46), 31719-31735 (2014-09-24)
Centrosome-mediated microtubule nucleation is essential for spindle assembly during mitosis. Although γ-tubulin complexes have primarily been implicated in the nucleation process, details of the underlying mechanisms remain poorly understood. Here, we demonstrated that a member of the human transforming acidic
Geun-Hyoung Ha et al.
Cancer letters, 332(1), 63-73 (2013-01-26)
Transforming acidic coiled-coil protein 3 (TACC3) is a member of the TACC family, essential for mitotic spindle dynamics and centrosome integrity during mitosis. Mounting evidence suggests that deregulation of TACC3 is associated with various types of human cancer. However, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service