Skip to Content
Merck
All Photos(3)

Documents

SAB4200360

Sigma-Aldrich

Anti-Cyclophilin B antibody, Mouse monoclonal

enhanced validation

clone CYPB-3, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-CYP-S1, Monoclonal Anti-CYPB, Monoclonal Anti-OI9, Monoclonal Anti-PPIB, Monoclonal Anti-SCYLP, Monoclonal Anti-peptidylprolyl isomerase B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

CYPB-3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~20 kDa

species reactivity

human, mouse, rat

enhanced validation

independent
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

western blot: 2.5-5.0 μg/mL using whole cell extracts of human HepG2 or rat NRK cells.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPIB(5479)
mouse ... Ppib(19035)
rat ... Ppib(64367)

General description

Cyclophilin B belongs to the cyclophilin family of immunophilins. It is present in the cytoplasm, nucleus, endoplasmic reticulum (ER)/golgi and plasma membrane. The protein can also be secreted from cells. The associated gene is mapped to human chromosome 15q22.
Monoclonal Anti-Cyclophilin B (mouse IgG1 isotype) is derived from the hybridoma CYPB-3 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to the C-terminal region of human Cyclophilin B, conjugated to keyhole limpet hemocyanin (KLH). Cyclophilin B (CypB) is a 21 kDa protein encoded by the peptidylprolyl isomerase B (PPIB) gene that belongs to the cyclophilin family, is a highly conserved class of proteins originally identified as cellular binding proteins for the immunosuppressive drug cyclosporin A. CypB is mainly located within the endoplasmic reticulum (ER) of all cell types.

Immunogen

synthetic peptide corresponding to the C-terminal region of human Cyclophilin B, conjugated to KLH. The corresponding sequence is identical in mouse, monkey and canine and differs by one amino acid in rat Cyclophilin B.

Application

Monoclonal Anti-Cyclophilin B antibody produced in mouse has been used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

Cyclophilin B (CypB) is involved in inflammation, viral infection and cancer. It also plays an important role in protecting cells against endoplasmic reticulum (ER) stress via its PPIase activity. Mutations in the PPIB gene give rise to recessive forms of osteogenesis imperfecta and knockdown of this gene decreases cell growth, proliferation and migration.
In endoplasmic reticulum (ER), Cyclophilin B works as a chaperone for exported proteins. It also participates in the intracellular transport of collagen and proper formation of collagen fibril. Cyclophilin B can bind to the Gag protein of human immunodeficiency type 1 (HIV-1) and helps in HIV replication. It is also involved in the infection of hepatitis C virus (HCV), Japanese encephalitis virus (JEV) and human papillomavirus type 16 (HPV16). Cyclophilin B protects against ER-mediated hypoxia-induced cell death. It is overexpressed in various malignancies.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hsp47 and Cyclophilin B Traverse the Endoplasmic Reticulum with Procollagen into Pre-Golgi Intermediate Vesicles A ROLE FOR Hsp47 AND CYCLOPHILIN B IN THE EXPORT OF PROCOLLAGEN FROM THE ENDOPLASMIC RETICULUM
Smith T, et al.
The Journal of Biological Chemistry, 270(31) (1995)
Chapter 12 - Radiology
Adams JE, et al.
Pediatric Bone (2012)
Expression of cyclophilin B is associated with malignant progression and regulation of genes implicated in the pathogenesis of breast cancer
Fang F, et al.
The American Journal of Pathology, 174(1), 297-308 (2009)
Cyclophilin B supports Myc and mutant p53-dependent survival of glioblastoma multiforme cells.
Choi JW, et al.
Cancer Research, 74, 484-484 (2014)
Cyclophilin B is involved in p300-mediated degradation of CHOP in tumor cell adaptation to hypoxia.
Jeong K, et al.
Cell Death and Differentiation, 21, 438-438 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service