Skip to Content
Merck
All Photos(1)

Key Documents

SAB3500106

Sigma-Aldrich

Anti-RUSC1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-NESCA, Anti-RUN and SH3 domain containing protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$491.00

$491.00


Estimated to ship on23 April 2025



Select a Size

Change View
100 μG
$491.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$491.00


Estimated to ship on23 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RUSC1(23623)

Immunogen

RUSC1 antibody was raised against a 17 amino acid peptide from near the carboxy terminus of human RUSC1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

RWDD3 (RSUME), a small RWD-containing protein, has a central role in sumoylation by enhancing SUMO conjugation in the regulatory network of immune–inflammatory signals. RWDD3 increases Ikbeta sumoylation and stability. In addition, RWDD3 inhibits TNF-alpha-induced kB-LUC (Luciferase) reporter activity, showing the functional consequence of Ikbeta increased stability. RSUME-enhanced sumoylation of Ikbeta leads to the inhibition of NF-kbeta activity on two well-known inflammatory genes, IL-8 and cyclooxygenase-2 (Cox-2) and therefore may also favor anti-inflammatory pathways. Expression of RWDD3 was induced under hypoxic conditions and it has a potential role during vascularization. Both BMP-4 and RWDD3 may be interesting targets for inhibiting steps involved in pituitary tumorigenesis.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500106.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service