Skip to Content
Merck
All Photos(2)

Key Documents

SAB1401793

Sigma-Aldrich

Monoclonal Anti-FKBP10 antibody produced in mouse

clone 1C6, ascites fluid

Synonym(s):

FKBP6, FKBP65, FLJ20683, FLJ22041, FLJ23833, hFKBP65

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$484.00

$484.00


Estimated to ship on26 April 2025



Select a Size

Change View
200 μL
$484.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$484.00


Estimated to ship on26 April 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1C6, monoclonal

species reactivity

human

technique(s)

capture ELISA: suitable
western blot: 1:500-1:1000

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... FKBP10(60681)

General description

The protein encoded by this gene belongs to the FKBP-type peptidyl-prolyl cis/trans isomerase family. It is located in endoplasmic reticulum and acts as molecular chaperones. An alternatively spliced variant encoding different isoform has been found, but the biological validity of the variant is not determined. (provided by RefSeq)

Immunogen

FKBP10 (NP_068758, 377 a.a. ~ 470 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
ADVVEIRTLSRPSETCNETTKLGDFVRYHYNCSLLDGTQLFTSHDYGAPQEATLGANKVIEGLDTGLQGMCVGERRQLIVPPHLAHGESGARGV

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Clear solution

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service