Skip to Content
Merck
All Photos(4)

Key Documents

SAB1401164

Sigma-Aldrich

Anti-GSTA4 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp686D21185, GSTA4-4, GTA4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$610.00

$610.00


Estimated to ship on27 April 2025



Select a Size

Change View
100 μG
$610.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$610.00


Estimated to ship on27 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GSTA4(2941)

General description

GSTA4 (glutathione S-transferase alpha 4) gene is mapped to human chromosome 6p12.2. GSTA4 belongs to the superfamily of detoxification enzymes and is expressed widely.

Immunogen

GSTA4 (NP_001503.1, 1 a.a. ~ 222 a.a) full-length human protein.

Sequence
MAARPKLHYPNGRGRMESVRWVLAAAGVEFDEEFLETKEQLYKLQDGNHLLFQQVPMVEIDGMKLVQTRSILHYIADKHNLFGKNLKERTLIDMYVEGTLDLLELLIMHPFLKPDDQQKEVVNMAQKAIIRYFPVFEKILRGHGQSFLVGNQLSLADVILLQTILALEEKIPNILSAFPFLQEYTVKLSNIPTIKRFLEPGSKKKPPPDEIYVRTVYNIFRP

Biochem/physiol Actions

Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. These enzymes are involved in cellular defense against toxic, carcinogenic, and pharmacologically active electrophilic compounds. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. This gene encodes a glutathione S-tranferase belonging to the alpha class. The alpha class genes, which are located in a cluster on chromosome 6, are highly related and encode enzymes with glutathione peroxidase activity that function in the detoxification of lipid peroxidation products. Reactive electrophiles produced by oxidative metabolism have been linked to a number of degenerative diseases including Parkinson′s disease, Alzheimer′s disease, cataract formation, and atherosclerosis. (provided by RefSeq)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leanne Ambrosio et al.
Antioxidants (Basel, Switzerland), 9(10) (2020-10-18)
The transcription factor nuclear factor erythroid 2-related factor 2 (Nrf2) is considered as the master regulator of antioxidant and cytoprotective gene expressions. Moreover, it plays a pivotal role in cancer progression. Nrf2 mediates the adaptive response which contributes to the
Down-regulation of glutatione S-transferase ? 4 (hGSTA4) in the muscle of thermally injured patients is indicative of susceptibility to bacterial infection.
Apidianakis Y
Faseb Journal, 26(2), 730-737 (2012)
Eric Ciamporcero et al.
Free radical biology & medicine, 115, 447-457 (2017-12-19)
Redox adaptation plays an important role in cancer cells drug resistance. The antioxidant response is principally mediated by the transcription factor Nrf2, that induces the transcriptional activation of several genes involved in GSH synthesis, chemoresistance, and cytoprotection. YAP is emerging
Martina Daga et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 56, 156-164 (2019-01-23)
Ailanthone (Aila) is a natural active compound isolated from the Ailanthus altissima, which has been shown to possess an "in vitro" growth-inhibitory effect against several cancer cell lines. Advanced bladder cancer is a common disease characterized by a frequent onset
Takehiro Matsumura et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 27(8), 3198-3208 (2013-05-08)
Steroidogenic factor 1 (SF-1) is a master regulator for steroidogenesis. In this study, we identified novel SF-1 target genes using a genome-wide promoter tiling array and a DNA microarray. SF-1 was found to regulate human glutathione S-transferase A (GSTA) family

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service