Skip to Content
Merck
All Photos(1)

Key Documents

SAB1103857

Sigma-Aldrich

Anti-MUSTN1 (6-20) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Musculoskeletal, embryonic nuclear protein 1, Anti-Mustang

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$473.00

$473.00


Estimated to ship on17 April 2025



Select a Size

Change View
200 μL
$473.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$473.00


Estimated to ship on17 April 2025


biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~9 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MUSTN1(389125)

General description

MUSTN1 gene was initially discovered in fracture callus tissue. The gene codes for a small nuclear protein and is known to be widely expressed in skeletal muscle. MUSTN1 expression is observed during embryogenesis in the developing muscles and somites. Its maximum expression is observed from the third month of life. The gene is highly conserved among the vertebrates. The gene is known to contain a nuclear localization sequence, phosphorylation and myristoylation sites.
Musculoskeletal, embryonic nuclear protein 1 (MUSTN1) is encoded by the gene is mapped to human chromosome 3p21.1.

Immunogen

synthetic peptide corresponding to amino acids 6-20of human MUSTN1

Biochem/physiol Actions

MUSTN1 is involved in bone development and regeneration, as well as chondrogenesis. It is critical in the regulation of myogenic differentiation and myofusion as its knock-down inhibits the processes.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert P Gersch et al.
Bone, 45(2), 330-338 (2009-05-05)
Mustn1 encodes a small nuclear protein expressed specifically in the musculoskeletal system that was originally identified as a strongly up-regulated gene during bone regeneration, especially in fracture callus proliferating chondrocytes. Further experiments were undertaken to investigate its expression and role
Allelic expression analysis of the osteoarthritis susceptibility locus that maps to chromosome 3p21 reveals cis-acting eQTLs at GNL3 and SPCS1
Gee F, et al.
BMC Medical Genetics, 15(1), 53-53 (2014)
Cheng Liu et al.
American journal of physiology. Cell physiology, 298(5), C1100-C1108 (2010-02-05)
Mustn1 (Mustang, musculoskeletal temporally activated novel gene) was originally identified in fracture callus tissue, but its greatest expression is detected in skeletal muscle. Thus, we conducted experiments to investigate the expression and function of Mustn1 during myogenesis. Temporally, quantitative real-time

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service