Skip to Content
Merck
All Photos(2)

Key Documents

PLA0114

Sigma-Aldrich

Rabbit anti-mTOR Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

FK506 binding protein 12-rapamycin associated protein 1, FK506 binding protein 12-rapamycin associated protein 2, FK506-binding protein 12-rapamycin complex-associated protein 1, FKBP-rapamycin associated protein, FKBP12-rapamycin complex-associated protein, FKBP12-rapamycin complex-associated protein 1, FLJ44809, FRAP, FRAP1, FRAP2, Mechanistic target of rapamycin, RAFT1, RAPT1, SKS, dJ576K7.1 (FK506 binding protein 12-rapamycin associated protein 1), mTOR, mammalian target of rapamycin, mechanistic target of rapamycin (serine/threonine kinase), rapamycin and FKBP12 target 1, rapamycin associated protein FRAP2, rapamycin target protein, rapamycin target protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$452.00

$452.00


Estimated to ship on26 April 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$452.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$452.00


Estimated to ship on26 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

immunoprecipitation (IP): 2-10 μg/mg
proximity ligation assay: suitable
western blot: 1:2,000-1:10,000

accession no.

NP_004949.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... mTOR(2475)

Immunogen

The epitope recognized by PLA0114 maps to a region between residue 200 and 250 of human Mammalian Target of Rapamycin using the numbering given in entry NP_004949.1 (GeneID 2475).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Studies of the mammalian target of rapamycin (mTOR) and its homologs demonstrate a role in integrating signals from growth factors, nutrients, stress, and cellular energy levels to control cell growth, translation initiation, ribosome biogenesis, and transcription factor localization. mTOR is the direct target of the cell cycle arresting activity of rapamycin. mTOR interacts with Raptor or Rictor to form the mTORC1 and mTORC2 complexes respectively. The mTORC1 complex also includes mLst8/GbetaL and functions to phosphorylate S6K and 4EBP1. The mTORC2 complex also includes mLst8/GbetaL, mSIN1 and protor-1 and functions to phosphorylate Akt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanjit K Dhar et al.
The Journal of biological chemistry, 294(17), 6831-6842 (2019-03-13)
Mitochondria are major sites of energy metabolism that influence numerous cellular events, including immunity and cancer development. Previously, we reported that the mitochondrion-specific antioxidant enzyme, manganese-containing superoxide dismutase (MnSOD), has dual roles in early- and late-carcinogenesis stages. However, how defective
Xiaoling Zhou et al.
Animal bioscience, 35(6), 847-857 (2022-01-07)
The effects of maternal undernutrition during midgestation on muscle fiber histology, myosin heavy chain (MyHC) expression, methylation modification of myogenic factors, and the mammalian target of rapamycin (mTOR) signaling pathway in the skeletal muscles of prenatal and postnatal goats were
Kaixuan Shi et al.
Cell death and differentiation, 30(1), 195-207 (2022-09-29)
Despite remarkable efficacy, targeted treatments often yield a subpopulation of residual tumor cells in part due to non-genetic adaptions. Previous mechanistic understanding on the emergence of these drug-tolerant persisters (DTPs) has been limited to epigenetic and transcriptional reprogramming. Here, by
Sanjit K Dhar et al.
Oncogene, 37(48), 6225-6242 (2018-07-25)
Autophagy is a highly regulated evolutionarily conserved metabolic process induced by stress and energy deprivation. Here, we show that DNA polymerase gamma (Polγ) deficiency activates a selective prosurvival autophagic response via mitochondria-mediated reactive oxygen species (ROS) signaling and the mammalian

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service