Skip to Content
Merck
All Photos(1)

Documents

P2736

Sigma-Aldrich

Pectinase from Aspergillus niger

aqueous solution

Synonym(s):

Pectinex 3XL®, Pectinex® 3X L

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

form

aqueous solution

concentration

≥3000 units/mL

storage temp.

2-8°C

Application

Petctinase is an enzyme from Aspergillus niger that is used in plant protoplast preparation to digest cell wall prior to organelle isolation. It has been used to conduct partial saccharification of sugars. Pectinases are used to study their role in the invasion of plant tissues by phytopathogens, the spoilage of produce and various food processing and plant biotechnology applications.

Biochem/physiol Actions

Pectolytic enzyme preparation produced from a selected strain of Aspergillus niger: contains mainly pectintranseliminase, polygalacturonase, and pectinesterase and small amounts of hemicellulases and cellulases. Pectinases hydrolyses pectin, which is a component of the cell wall. They may attack methyl-esterified pectin or de-esterified pectin. It is a source of pectinase activity, also containing cellulase and hemicellulase activities.

Legal Information

A product of Novozyme Corp.
Pectinex is a registered trademark of Novozymes Corp.

replaced by

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Meredith C Edwards et al.
Applied and environmental microbiology, 77(15), 5184-5191 (2011-06-15)
Ethanologenic Escherichia coli strain KO11 was sequentially engineered to contain the Klebsiella oxytoca cellobiose phosphotransferase genes (casAB) as well as a pectate lyase (pelE) from Erwinia chrysanthemi, yielding strains LY40A (casAB) and JP07 (casAB pelE), respectively. To obtain an effective
Sara Posé et al.
Journal of experimental botany, 64(12), 3803-3815 (2013-07-23)
Antisense-mediated down-regulation of the fruit-specific polygalacturonase (PG) gene FaPG1 in strawberries (Fragaria×ananassa Duch.) has been previously demonstrated to reduce fruit softening and to extend post-harvest shelf life, despite the low PG activity detected in this fruit. The improved fruit traits
Isabelle Benoit et al.
BMC genomics, 13, 321-321 (2012-07-21)
Pectins are diverse and very complex biomolecules and their structure depends on the plant species and tissue. It was previously shown that derivatives of pectic polymers and oligosaccharides from pectins have positive effects on human health. To obtain specific pectic
Joan Ho-Huu et al.
BMC evolutionary biology, 12, 195-195 (2012-10-03)
Gene duplications are a molecular mechanism potentially mediating generation of functional novelty. However, the probabilities of maintenance and functional divergence of duplicated genes are shaped by selective pressures acting on gene copies immediately after the duplication event. The ratio of
Ding-Tao Wu et al.
Carbohydrate polymers, 97(2), 398-405 (2013-08-06)
Polysaccharides from Ganoderma spp. and their adulterants were firstly investigated and compared using saccharide mapping, enzymatic (endo-1,3-β-D-glucanase and pectinase) digestion followed by polysaccharide analysis using carbohydrate gel electrophoresis analysis. The results showed that both 1,3-β-D-glucosidic and 1,4-α-D-galactosiduronic linkages were existed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service