Skip to Content
Merck
All Photos(3)

Key Documents

MAK069

Sigma-Aldrich

MPO Fluorometric Activity Assay Kit

sufficient for 100 fluorometric tests

Synonym(s):

Myeoloperoxidase Fluorometric Activity Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503

usage

sufficient for 100 fluorometric tests

detection method

fluorometric

relevant disease(s)

cancer; cardiovascular diseases; hematological disorder

storage temp.

−20°C

Gene Information

human ... MPO(4353)
mouse ... MPO(17523)
rat ... MPO(303413)

General description

Myeloperoxidase (MPO) is a heme-containing enzyme that catalyzes the hydrogen peroxidase-mediated oxidation of halide ions to hypohalous acid. MPO is a lysosomal protein, highly expressed in neutrophils, that plays a role in the antimicrobial actions that occur as a result of neutrophil stimulation. The actions of MPO may also contribute to the initiation and pathogenesis of cardiovascular disease.

Application

MPO Fluorometric Activity Assay Kit has been used to determine the activity of myeloperoxidase.

Suitability

Suitable for the measurement of myeloperoxidase (MPO) in biological samples such as tissue, cells, serum and white blood cells

Principle

The MPO Activity Flurometric Assay kit provides a simple and direct procedure for measuring MPO activity in a variety of samples. In this assay, MPO catalyzes the formation of hypochlorous acid with reacts with the substrate, aminophenyl fluorescein, to generate fluorescein) (λex = 485/λem = 525 nm). One unit of MPO is the amount of enzyme that will oxidize the MPO substrate to yield 1.0 μmole of fluorescein per minute at room temperature.

replaced by

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minghui Li et al.
International journal of cardiology, 221, 1123-1131 (2016-07-30)
Viral myocarditis (VMC) treatment has long been lacking of effective methods. Our former studies indicated roles of calpain in VMC pathogenesis. This study aimed at verifying the potential of calpain in Coxsackievirus B3 (CVB3)-induced myocarditis treatment. A transgenic mouse overexpressing
Harish Chandra Pal et al.
Photochemistry and photobiology, 91(1), 225-234 (2014-08-30)
Solar ultraviolet B (UVB) radiation has been shown to induce inflammation, DNA damage, p53 mutations and alterations in signaling pathways eventually leading to skin cancer. In this study, we investigated whether fisetin reduces inflammatory responses and modulates PI3K/AKT/NFκB cell survival
Macrophage micro-RNA-155 promotes lipopolysaccharide-induced acute lung injury in mice and rats.
Wang W, et al.
American Journal of Physiology. Lung Cellular and Molecular Physiology, 311(2), L494-L506 (2016)
Wen Wang et al.
American journal of physiology. Lung cellular and molecular physiology, 311(2), L494-L506 (2016-07-03)
Micro-RNA (miR)-155 is a novel gene regulator with important roles in inflammation. Herein, our study aimed to explore the role of miR-155 in LPS-induced acute lung injury(ALI). ALI in mice was induced by intratracheally delivered LPS. Loss-of-function experiments performed on
Ali Skalli et al.
Animals : an open access journal from MDPI, 10(9) (2020-09-19)
A nutritional study was conducted to evaluate the inclusion of the green microalga Scenedesmus sp. at 5% (SCE-5) as an alternative fishmeal ingredient. This microalga was tested with four replicates during 45 days using isolipidic (18%), isoproteic (48%), and isoenergetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service