Skip to Content
Merck
All Photos(1)

Documents

K1127

Sigma-Aldrich

Kemptide acetate salt

≥95% (HPLC), powder

Synonym(s):

Phosphate acceptor peptide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H61N13O9 · xC2H4O2
CAS Number:
Molecular Weight:
771.91 (free base basis)
Beilstein:
6263936
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95% (HPLC)

form

powder

color

white

solubility

H2O: 20 mg/mL

storage temp.

−20°C

SMILES string

CC(O)=O.[H]N[C@@H](CC(C)C)C(N[C@@H](CCCNC(N)=N)C(N[C@@H](CCCNC(N)=N)C(N[C@H](C(N[C@@H](CO)C(N[C@@H](CC(C)C)C(NCC(O)=O)=O)=O)=O)C)=O)=O)=O

InChI

1S/C32H61N13O9.C2H4O2/c1-16(2)12-19(33)26(50)42-21(9-7-11-39-32(36)37)29(53)43-20(8-6-10-38-31(34)35)28(52)41-18(5)25(49)45-23(15-46)30(54)44-22(13-17(3)4)27(51)40-14-24(47)48;1-2(3)4/h16-23,46H,6-15,33H2,1-5H3,(H,40,51)(H,41,52)(H,42,50)(H,43,53)(H,44,54)(H,45,49)(H,47,48)(H4,34,35,38)(H4,36,37,39);1H3,(H,3,4)/t18-,19-,20-,21-,22-,23-;/m0./s1

InChI key

WRTDQEIEUYKAKV-PXILYFGCSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Tortora et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(5), 2011-2015 (1991-03-01)
A marked decrease in the type I cAMP-dependent protein kinase regulatory subunit (RI alpha) and an increase in the type II protein kinase regulatory subunit (RII beta) correlate with growth inhibition and differentiation induced in a variety of types of
J L Maller et al.
Proceedings of the National Academy of Sciences of the United States of America, 75(1), 248-251 (1978-01-01)
A model synthetic peptide substrate of the cyclic AMP-dependent protein kinase (ATP:protein phosphotransferase; EC 2.7.1.37), Leu-Arg-Arg-Ala-Ser-Leu-Gly, closely resembling the local phosphorylation site sequence in porcine hepatic pyruvate kinase, was shown to be phosphorylated in vivo after microinjection into Xenopus oocytes.
N-terminal protein sequencing for special applications.
Philip J Jackson
Methods in molecular biology (Clifton, N.J.), 211, 287-300 (2002-12-20)
Role of multiple basic residues in determining the substrate specificity of cyclic AMP-dependent protein kinase.
B E Kemp et al.
The Journal of biological chemistry, 252(14), 4888-4894 (1977-07-25)
L Belloni-Olivi et al.
The Biochemical journal, 315 ( Pt 2), 401-406 (1996-04-15)
In immature rat microvessels, endothelial cells and glioma cells, exposure to lead results in an increase in the level of protein kinase C in membranes. In this paper we have extended these studies to human erythrocytes and, in addition, studied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service