Skip to Content
Merck
All Photos(4)

Key Documents

HPA018121

Sigma-Aldrich

Anti-BAG1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BAG family molecular chaperone regulator 1, Anti-BAG-1, Anti-Bcl-2-associated athanogene 1, Anti-Glucocorticoid receptor-associated protein RAP46

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$952.00

$952.00


Estimated to ship on20 April 2025



Select a Size

Change View
100 μL
$952.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$952.00


Estimated to ship on20 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

GKSLKEMETPLSALGIQDGCRVMLIGKKNSPQEEVELKKLKHLEKSVEKIADQLEELNKELTGIQQGFLPKDLQAEALCKLDRRVKATIEQFMKILEEIDTLILPENFKDSRLKRKGLVKKVQAFLAECDTVEQNICQETE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BAG1(573)

General description

BCL2 associated athanogene 1 (BAG1) is an anti-apoptotic protein and the gene expressing it is localized on human chromosome 9.

Immunogen

BAG family molecular chaperone regulator 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

BCL2 associated athanogene 1 (BAG1) has been shown to be upregulated in cancers. It binds to B-cell lymphoma 2 (BCL-2) and other proteins. BAG1 modulates various signal transduction pathways and also regulates apoptosis.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72797

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Liu et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 33(2), 365-374 (2014-02-22)
BAG-1 (bcl-2 associated athanogene) is a multifunctional protein that protects cells from a wide range of apoptotic stimuli including radiation, hypoxia and chemotherapeutic agents. Overexpression of cytoplasmic BAG-1 has been associated with the increased survival and decreased response to treatment
Bo Huang et al.
Oncology reports, 32(4), 1441-1446 (2014-07-30)
The aim of the present study was to explore the correlation of BAG-1 with clinical characteristics of esophageal cancer and its effects on the proliferation, invasion and apoptosis of the esophageal carcinoma cell line Eca109. Therefore, the expression of BAG-1
Maria Afentakis et al.
Breast cancer research and treatment, 140(2), 253-262 (2013-07-16)
BAG1 is a multifunctional anti-apoptotic protein located on chromosome 9q12, which binds to Bcl-2. BAG1 is present as a separate module in the GHI-RS 21-gene panel. It may provide additional prognostic information as an immunohistochemical marker when added to IHC4.
Fei Ma et al.
PloS one, 10(5), e0126499-e0126499 (2015-05-12)
MiR-138 is frequently downregulated in different cancer types and is thought to be involved in the progression of tumorigenesis. However, the molecular mechanism of miR-138 involvement in gallbladder carcinoma still remains unknown. The expression of miR-138 in 49 gallbladder carcinoma

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service