Skip to Content
Merck
All Photos(2)

Documents

HPA015242

Sigma-Aldrich

Anti-ERGIC3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Endoplasmic reticulum-Golgi intermediate compartment protein 3, Anti-Serologically defined breast cancer antigen NY-BR-84

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

CNTCEDVREAYRRRGWAFKNPDTIEQCRREGFSQKMQEQKNEGCQVYGFLEVNKVAGNFHFAPGKSFQQSHVH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ERGIC3(51614)

General description

ERGIC3 (Endoplasmic reticulum-Golgi intermediate compartment protein 3) is a membrane protein that shuttles between the Golgi apparatus and the endoplasmic reticulum (ER). It has large domains which face the ER lumen, and the small N- and C-termini face the cytoplasm. This protein is composed of 383 amino acids, and has a molecular weight of 43kDa. It has a high level of expression in kidney and the spinal cord. This gene is localized to human chromosome 20.

Immunogen

Endoplasmic reticulum-Golgi intermediate compartment protein 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ERGIC3 (Endoplasmic reticulum-Golgi intermediate compartment protein 3) is responsible for the trafficking of proteins from the endoplasmic reticulum (ER) to the Golgi apparatus. As it co-localizes with MUC1 (Mucin 1) and ST proteins, it might be involved in their trafficking as well. It is also involved in ER-stress related apoptosis and cellular growth. It is overexpressed in lung cancer cells, and this is correlated to the histological grade and the differentiation of the lung cancer tissue. Thus, this gene might be linked to cancer proliferation and metastasis. miR-490-3p is a positive regulator of ERGIC3, which leads to enhanced tumor proliferation and metastasis of hepatocellular carcinoma cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72866

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manabu Nishikawa et al.
Gene, 386(1-2), 42-51 (2006-10-06)
Disposal of misfolded proteins from the lumen of the endoplasmic reticulum (ER) is one of the quality control mechanisms present in the protein secretory pathway. Through ER-associated degradation, misfolded substrates are targeted to the cytosol where they are degraded by
Ling-yun Zhang et al.
The Journal of biological chemistry, 288(6), 4035-4047 (2012-12-06)
MicroRNAs (miRNAs) are considered to be regulators of various biological processes in cancers, including the epithelial to mesenchymal transition (EMT), which is a key factor in cancer metastasis. In this study, we aimed to clarify the potential roles of miR-490-3p
Mingsong Wu et al.
BMC cancer, 13, 44-44 (2013-02-05)
To understand the carcinogenesis caused by accumulated genetic and epigenetic alterations and seek novel biomarkers for various cancers, studying differentially expressed genes between cancerous and normal tissues is crucial. In the study, two cDNA libraries of lung cancer were constructed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service