Skip to Content
Merck
All Photos(1)

Key Documents

HPA005442

Sigma-Aldrich

Anti-KIF1A antibody produced in rabbit

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ATSV, Anti-C2orf20, Anti-SPG30, Anti-UNC104

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

SGTAKISFDDQHFEKFQSESCPVVGMSRSGTSQEELRIVEGQGQGADVGPSADEVNNNTCSAVPPEGLLLDSSEKAALDGPLDAALDHLRLGNTFTFRVTVLQASSISAEYADIF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... KIF1A(547)

Looking for similar products? Visit Product Comparison Guide

General description

Kinesin Family (KIF) of proteins is responsible for the intracellular transport of mRNAs, membrane vesicles, organelles and protein complexes, in polarized cells. They are molecular motors, which are microtubule dependent. They are divided into 14 subfamilies through kinesin-1 to kinesin-14. KIF1A protein is a part of kinesin-3 subfamily and is composed of 1690 amino acids. KIF1A is an N-kinesin as its motor domain is present at the N-terminal.

Immunogen

Kinesin-like protein KIF1A recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Kinesin Family 1A (KIF1A) protein drives transport in an anterograde manner, i.e. the microtubule plus end towards the axon terminal. It transports the synaptic vesicle precursors to the axon terminal. The precursors contain synaptic vesicle proteins, synaptophysin, synaptotagmin, as well as the GTPase RAB3A. RAB3 then causes the exocytosis of neurotransmitters contained in the synaptic vesicles at the axon end. During brain development in embryos, KIF1A regulates the interkinetic nuclear migration. Two autosomal recessive families of hereditary spastic paraplegias (HSPs) have been linked to mutations in KIF1A gene. Also, mutations in KIF1A gene are a rare cause of hereditary sensory and autonomic neuropathy type 2 (HSANII).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86829.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephan Klebe et al.
European journal of human genetics : EJHG, 20(6), 645-649 (2012-01-20)
The hereditary spastic paraplegias (HSPs) are a clinically and genetically heterogeneous group of neurodegenerative diseases characterised by progressive spasticity in the lower limbs. The nosology of autosomal recessive forms is complex as most mapped loci have been identified in only
Julian Scherer et al.
PLoS pathogens, 16(1), e1007985-e1007985 (2020-01-30)
Axonal sorting, the controlled passage of specific cargoes from the cell soma into the axon compartment, is critical for establishing and maintaining the polarity of mature neurons. To delineate axonal sorting events, we took advantage of two neuroinvasive alpha-herpesviruses. Human
Yaniv Erlich et al.
Genome research, 21(5), 658-664 (2011-04-14)
Whole exome sequencing has become a pivotal methodology for rapid and cost-effective detection of pathogenic variations in Mendelian disorders. A major challenge of this approach is determining the causative mutation from a substantial number of bystander variations that do not
Jean-Baptiste Rivière et al.
American journal of human genetics, 89(2), 219-230 (2011-08-09)
Hereditary sensory and autonomic neuropathy type II (HSANII) is a rare autosomal-recessive disorder characterized by peripheral nerve degeneration resulting in a severe distal sensory loss. Although mutations in FAM134B and the HSN2 exon of WNK1 were associated with HSANII, the
Lin Huo et al.
Structure (London, England : 1993), 20(9), 1550-1561 (2012-08-07)
Kinesin-3 KIF1A plays prominent roles in axonal transport and synaptogenesis. KIF1A adopts a monomeric form in vitro but acts as a processive dimer in vivo. The mechanism underlying the motor dimerization is poorly understood. Here, we find that the CC1-FHA tandem of KIF1A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service