Skip to Content
Merck
All Photos(1)

Documents

F1513

Sigma-Aldrich

Iron(III) chloride hexahydrate

ACS reagent, 97.0-102.0%

Synonym(s):

Ferric chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl3 · 6H2O
CAS Number:
Molecular Weight:
270.30
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

ACS reagent

vapor pressure

1 mmHg ( 194 °C)

Assay

97.0-102.0%

impurities

≤0.01% Insoluble matter
≤0.01% Phosphorus compounds (as PO4)
≤0.1% Substances not precipitated by ammonium hydroxide (as sulfates)

bp

280-285 °C (lit.)

mp

37 °C (lit.)

anion traces

nitrate (NO3-): ≤0.01%
sulfate (SO42-): ≤0.01%

cation traces

Cu: ≤0.003%
Fe2+:, passes test (limit ∼0.002%)
Zn: ≤0.003%

SMILES string

O.O.O.O.O.O.Cl[Fe](Cl)Cl

InChI

1S/3ClH.Fe.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

NQXWGWZJXJUMQB-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanjay Kashyap et al.
ACS nano, 8(9), 9097-9106 (2014-08-28)
Biomineralization proteins are widely used as templating agents in biomimetic synthesis of a variety of organic-inorganic nanostructures. However, the role of the protein in controlling the nucleation and growth of biomimetic particles is not well understood, because the mechanism of
D C Florian Wieland et al.
Colloids and surfaces. B, Biointerfaces, 109, 74-81 (2013-04-27)
The early stages of the formation of inorganic aggregates, composed of iron compounds at the solution-air interface, were investigated in situ. The properties of the solution-air interface were changed by using different Langmuir layers. In order to get insight into
Dattatraya H Dethe et al.
Organic letters, 15(3), 429-431 (2013-01-16)
A novel approach was developed for the synthesis of highly substituted indene derivatives, using an FeCl(3) catalyzed Prins-type cyclization reaction which was further applied in the total synthesis of jungianol and epi-jungianol.
Justin D Barr et al.
Blood, 121(18), 3733-3741 (2013-01-25)
Application of ferric chloride (FeCl(3)) to exposed blood vessels is widely used to initiate thrombosis in laboratory mice. Because the mechanisms by which FeCl(3) induces endothelial injury and subsequent thrombus formation are little understood, we used scanning electron and brightfield
Xiaoqing Li et al.
The Journal of organic chemistry, 78(18), 9499-9504 (2013-08-28)
Halosulfonylation of terminal alkynes was achieved with sulfonylhydrazides as the sulfonyl precursor and inexpensive iron halide as halide source in the presence of TBHP, allowing the regio- and stereoselective generation of (E)-β-chloro and bromo vinylsulfones.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service