Skip to Content
Merck
All Photos(1)

Key Documents

EPS44I040006

Eppendorf® Dedicated Platform

for flasks 2000 mL, Innova(R) S44i

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103800
Product EPS44I040006 is not currently sold in your country. Contact Technical Service

feature

for flasks 2000 mL, Innova(R) S44i

manufacturer/tradename

S44I040006
Eppendorf®

Looking for similar products? Visit Product Comparison Guide

General description

Dedicated ppendorf shelves to hold various volume size flasks securely while shaking in Innova S44i.
Dedicated shelf for the Innova S44i. Holds up to 15 x 2 L Erlenmeyer flasks.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service