EP5920710001
Adapter for Eppendorf® S-4x1000 Rotor
capacity, 22 x Eppendorf Tube® 5.0 mL, for use with medium-capacity buckets, pack of 2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
capacity 22 x Eppendorf Tube® 5.0 mL
packaging
pack of 2
manufacturer/tradename
Eppendorf® 5920710001 (Global)
parameter
3,076 × g max. RCF
technique(s)
centrifugation: suitable
H
121 mm
diam.
17 mm
compatibility
for use with plate/tube bucket
Looking for similar products? Visit Product Comparison Guide
General description
Adapters to fit various size tubes and bottles for use with Eppendorf S-4x1000 rotor.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Tube is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service