Skip to Content
Merck
All Photos(1)

Key Documents

EHU113611

Sigma-Aldrich

MISSION® esiRNA

targeting human FOXO3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TATGCAAACCCTCTCGGACTCTCTCTCAGGCTCCTCCTTGTACTCAACTAGTGCAAACCTGCCCGTCATGGGCCATGAGAAGTTCCCCAGCGACTTGGACCTGGACATGTTCAATGGGAGCTTGGAATGTGACATGGAGTCCATTATCCGTAGTGAACTCATGGATGCTGATGGGTTGGATTTTAACTTTGATTCCCTCATCTCCACACAGAATGTTGTTGGTTTGAACGTGGGGAACTTCACTGGTGCTAAGCAGGCCTCATCTCAGAGCTGGGTGCCAGGCTGAAGGATCACTGAGGAAGGGGAAGTGGGCAAAGCAGACCCTCAAACTGACACAAGACCTACAGAGAAAACCCTTTGCCAAATCTGCTCTCAGCAAGTGGACAGTGATACCGTTTACAGCTTAACACCTTTGTGAATCCCACGCCATTTTCCTAACCCAGCAGAGACTGTTAATGGCCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ning Liu et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 66(7), 603-610 (2017-04-13)
Fibroblast-like synoviocytes (FLS) play an essential role in the pathogenesis of chronic inflammatory diseases, such as rheumatoid arthritis. Paeonol (Pae) is a phenolic compound found in many traditional Chinese medicine remedies. However, the effects of Pae on TNF-α-stimulated FLS and
M Kumazoe et al.
Oncogene, 36(19), 2643-2654 (2016-11-29)
Pancreatic ductal adenocarcinoma (PDAC) is one of the most fatal types of cancer and the 5-year survival rate is only 5%. Several studies have suggested that cancer stem cells (CSCs) are thought to be involved in recurrence and metastasis and
Yanqiu Wang et al.
Biochemical and biophysical research communications, 524(3), 756-763 (2020-02-10)
Intervertebral disc degeneration (IDD) is typically accompanied by a reduced nutrient supply, which is thought to be a contributor to the apoptosis of nucleus pulposus cells (NPCs). Here, we explored whether Forkhead box O3 (FOXO3), a key transcription factor involved
Fei Wang et al.
Folia histochemica et cytobiologica, 58(1), 1-8 (2020-02-01)
Osteoarthritis (OA) is the most common degenerative disease in middle-aged and elderly individuals that causes joint deformity and limb disability. Accumulating evidence has suggested that the pathogenesis of OA has been related to various mechanisms such as apoptosis, inflammation, oxidative
Huaping Zhang et al.
Toxicology letters, 277, 115-122 (2017-07-10)
The antioxidant activity of genistein is associated with preventing atherosclerosis; however, the underlying mechanisms are not fully understood. In this study, human umbilical vein endothelial cells (HUVECs) were pretreated with genistein at different concentrations (10nM, 100nM and 1000nM) for 6h

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service