Skip to Content
Merck
All Photos(1)

Documents

D6071

Sigma-Aldrich

Diphenylacetohydroxamic acid

≥98% (HPLC)

Synonym(s):

N-Hydroxy diphenylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13NO2
CAS Number:
Molecular Weight:
227.26
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

off-white to light tan

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

ONC(=O)C(c1ccccc1)c2ccccc2

InChI

1S/C14H13NO2/c16-14(15-17)13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,13,17H,(H,15,16)

InChI key

OFSBBVCLWMCGNY-UHFFFAOYSA-N

Biochem/physiol Actions

Diphenylacetohydroxamic acid is a class IIa selective histone deacetylase inhibitor.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Douglas D Lemon et al.
FEBS letters, 589(10), 1080-1088 (2015-03-31)
PKD-mediated phosphorylation of class IIa HDACs frees the MEF2 transcription factor to activate genes that govern muscle differentiation and growth. Studies of the regulation and function of this signaling axis have involved MC1568 and Gö-6976, which are small molecule inhibitors

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service