Skip to Content
Merck
All Photos(1)

Key Documents

COLO 320DM

87061205, human colon, Round & Refractile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

COLO 320DM, 87061205

biological source

human colon

growth mode

Suspension

karyotype

Hyperdiploid, modal no. 53

morphology

Round & Refractile

products

Serotonin, epinephrine, NPP, Adrenocorticotropic hormone (ACTH) and PTH

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian colon adenocarcinoma

Cell Line Description

Isolated from a moderately undifferentiated adenocarcinoma from the sigmoid colon of a 55 year old Caucasian female. The cells have double minute (DM) chromosomes.

Application

Hormone studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11,12
D13S317: 11
D16S539: 11,12
D5S818: 12
D7S820: 9,12
THO1: 8,9
TPOX: 8,9
vWA: 15,18

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. may semi-adhere to culture flask.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service