Skip to Content
Merck
All Photos(1)

Documents

C9783

Sigma-Aldrich

Z-His(Bzl)-OH

≥98% (TLC)

Synonym(s):

Nα-Cbz-Nim-benzyl-L-histidine, Nα-Z-N(im)-benzyl-L-histidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21N3O4
CAS Number:
Molecular Weight:
379.41
Beilstein:
55593
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

product name

Z-His(Bzl)-OH,

Assay

≥98% (TLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

OC(=O)[C@H](Cc1ccn(Cc2ccccc2)n1)NC(=O)OCc3ccccc3

InChI

1S/C21H21N3O4/c25-20(26)19(22-21(27)28-15-17-9-5-2-6-10-17)13-18-11-12-24(23-18)14-16-7-3-1-4-8-16/h1-12,19H,13-15H2,(H,22,27)(H,25,26)/t19-/m0/s1

InChI key

DHUVTQUENUPYMW-IBGZPJMESA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Z-His(Bzl)-OH (Nα-Cbz-Nim-benzyl-L-histidine) is an N-terminal blocked (with CBZ) benzolated (on Imidazole) L-histidine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service